Microcentrifuge tube opener, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F199250001
BELAF199250001EA 71.1 EUR InStock
BELAF199250001
Microcentrifuge tube opener, SP Bel-Art
Crimper, Capper und Decapper
Easily open all sizes of microcentrifuge tubes with this double-ended tool.
  • Opens standard 1,5, 2,0 and 2,2 ml tubes with one end
  • Forked blade on other end opens smaller sizes (250, 400 and 550 μl)
  • Dimensions: 7 cm (3⅛") long
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge