Spill containment tray with grid, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F246760003
BELAF246760003EA 256 EUR InStock
BELAF246760003
Spill containment tray with grid, SP Bel-Art
Schalen/Schüsseln/Töpfe Auffangschalen und Auffangbehälter
This large, deep, polypropylene plastic tray protects users by capturing accidental hazardous spills.
  • High chemical resistance
  • Epoxy coated wire support grid holds numerous vessels or racks of test tubes
  • Steam autoclavable at 121 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge