Glove box holders, Poxygrid®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F247390002 F247390003 F247390001
BELAF247390002EA 154 EUR InStock
BELAF247390002 BELAF247390003 BELAF247390001
Glove box holders, Poxygrid®, SP Bel-Art
Halterungen für Handschuhspender und -boxen
Epoxy-coated, welded steel wire racks for one, two or three boxes of disposable gloves, wipes or tissues.
  • Glove box drops into an individual retaining compartment and lifts out easily for replacement
  • Designed for mounting to a vertical surface
  • Durable, blue epoxy coated racks are steam autoclavable at 121 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge