Scoops, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F367530000 F367560000 F367480000 F367490000 F367470000 F367500000
231-0397EA 84.8 EUR InStock
231-0397 231-0398 BELAF367480000 231-2419 BELAF367470000 BELAF367500000
Scoops, SP Bel-Art
Schaufeln
PP, translucent. These plastic scoops are rigid, strong and smooth. A flat bottom keeps them upright when filled or empty, allowing them to be used as weighing boats or to be placed on a flat surface.
  • 60 and 125 ml scoops have a longer handle that increases the distance between the hand and sample
  • Rugged PP; steam autoclavable at 121 °C
  • Comply with FDA regulations for food contact
The sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge