Scraper, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F368200001
BELAF368200001EA 90.9 EUR InStock
BELAF368200001
Scraper, SP Bel-Art
Schaber
PP, white.
  • Rigid, chisel-edged, triangular blade is tapered and fans out to a width of 11,1 cm (4⅜")
  • Handle is shaped for a comfortable and secure grip
  • Steam autoclavable at 121 °C
  • Complies with FDA requirements for direct contact with foods, drugs and cosmetics
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge