Cub - dry ice maker, Frigimat®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F388740000
BELAF388740000EA 1550 EUR InStock
BELAF388740000
Cub - dry ice maker, Frigimat®, SP Bel-Art
Eisbereiter
This compact device can be attached to a liquid CO₂ cylinder (not included) equipped with a dip tube to produce a ready-to-use 250 to 350 g block of dry ice in only a few minutes.
  • Lightweight body allows safe transport of the unit and contents using appropriate gloves
  • Operates at very low pressure
  • Rugged nickel-plated 'pigtail' connects it to the CO₂ cylinder valve which is used to regulate gas flow
Dry ice production can be viewed through the transparent polycarbonate body that secures without clamps, screws or tools ensuring easy removal of the solid block of dry ice and disconnection from the CO₂ cylinder.
Lieferumfang: It includes nickel-plated copper connecting tube (1,8 m) with standard CGA 320 thread coupling for easy attachment to a CO₂ cylinder.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge