Dispensing jug, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H118500000
BELAH118500000EA 162 EUR InStock
BELAH118500000
Dispensing jug, SP Bel-Art
Flaschen Ballons
LDPE, translucent.
  • Strong and lightweight
  • Storage capacity is 20 L (5 gallons)
  • Gradations in gallons are moulded into the side
Use this jug to store and dispense common solutions, including janitorial cleaners, deionized/distilled water, or other aqueous liquids. Threaded LDPE spigot extends 125 mm from jug and features a 35 mm long stem for dispensing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge