Pulvertrichter
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H146600080 H146600065 H146600100 H146600150
BELAH146600080EA 170 EUR InStock
BELAH146600080 BELAH146600065 BELAH146600100 BELAH146600150
Pulvertrichter
Trichter Pulvertrichter
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
Specially designed for transferring powders. These funnels have 60º angle and a wide stem for clog-free filling.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge