Electrode rack, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H185090000
BELAH185090000EA 243 EUR InStock
BELAH185090000
Electrode rack, SP Bel-Art
Elektrodenhalter
This handy rack made from PP holds eight test tubes for storing and conditioning pH and ion electrodes.
  • Holds two rows of four tubes up to 20 mm in diameter
  • PP rack resists chemical attack
  • Autoclavable at 121 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge