Separating funnel stands, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H189640000
BELAH189640000EA 588 EUR InStock
BELAH189640000
Separating funnel stands, SP Bel-Art
Stative
This rotary separatory funnel rack consists of a rectangular base, screw-in column and a support disc held in place by a screw clamp.
  • Support disc is fully adjustable for different heights up to 21"
  • Open front type support holes for easy removal and placement of separatory funnels
  • Shipped unassembled
Bestellinformation: Includes 18 insert clips to accommodate different sized funnels.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge