Disposable paper sampling spoons, EcoTensil®
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H367401040
BELAH367401040EA 0 EUR InStock
BELAH367401040
Disposable paper sampling spoons, EcoTensil®
Probenehmer Probenehmer (Einweg)
This paperboard product stores flat and easily folds into a functional spoon for sampling and retrieving materials in the laboratory.
  • Coated to stand up to powders, gels, and semi-solids
  • Use 50% - 90% less material and space than the same amount of plastic spoons
  • Compostable (ASTM D6868) and recyclable at facilities where paper coffee cups are accepted
  • Low cost way to go green in the laboratory
  • Can be used as a weigh boat eliminating the need for glassine sheets
  • Made in the USA
NOTE: EcoTensil® Spoons are not meant for measuring exact quantities of materials. The capacity given in the ordering table is the approximate capacity.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge