Mini mortar with pestle, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H372600100
BELAH372600100EA 1570 EUR InStock
BELAH372600100
Mini mortar with pestle, SP Bel-Art
Mörser und Pistille Mörser
Freeze and process small tissue samples within a microcentrifuge tube. Stainless steel mortar has a welded fixture to hold a 1,5ml microcentrifuge tube. The mortar is suspended above a stainless steel cavity which holds the liquid nitrogen and is nested in a heavily insulated blue polyethylene housing.
  • Prevents tube from cracking by eliminating direct contact with liquid nitrogen
  • Includes 12,7mm long stainless steel pestle with 102×19mm polyethylene handle
  • Suction feet keep it stable on the workbench
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge