Guarded flow meters, Riteflow®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H404030235
BELAH404030235EA 929 EUR InStock
BELAH404030235
Guarded flow meters, Riteflow®, SP Bel-Art
Durchflussmesser
These flow meters has a plastic safety guard over the flowtube.
  • Heavy-walled shock-resistant borosilicate glass
  • Fused and enclosed ceramic scales
  • Uniformly tapered bores with internal rib guides
  • Floats are retained by Teflon® plugs
Used for flow measurements in the lab, process flows in pilot plants, metering carrier gases in chromatography, indicating and controlling gases in manufacturing processes, and for indicating flow rates in pumping or flow systems.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge