Pipette canisters, round bottom
Lieferant: BELLCO GLASS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1287-00430 1287-38250 1287-38430
BELC1287-00430EA 475 EUR InStock
BELC1287-00430 BELC1287-38250 BELC1287-38430
Pipette canisters, round bottom
Dosen Pipettenbehälter
Pipette canister made of borosilicate glass, class A, type I.
  • Manufactured from Type 1, Class A borosilicate glass
  • Components will withstand repeated dry or steam heat sterilisation
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge