Pipette canisters, flat bottom
Lieferant: BELLCO GLASS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1290-00250 1290-38250 1290-38430 1290-00300
BELC1290-00250EA 247 EUR InStock
BELC1290-00250 BELC1290-38250 BELC1290-38430 BELC1290-00300
Pipette canisters, flat bottom
Dosen Pipettenbehälter
Glass pipette canisters constructed of borosilicate glass.
  • With flat bottom
  • All components will withstand repeated dry or steam heat sterilization
  • Bottom silicone pad prevents chipping of pipette tips
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge