Microcarrier spinner flasks
Lieferant: BELLCO GLASS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1965-00100 1965-00250 1965-00500 1965-01000 1965-03000 1965-06000 1965-15000 1965-02500 1965-08000
BELC1965-00100EA 550 EUR InStock
BELC1965-00100 BELC1965-00250 BELC1965-00500 BELC1965-01000 BELC1965-03000 BELC1965-06000 BELC1965-15000 BELC1965-02500 BELC1965-08000
Microcarrier spinner flasks
Kolben Spinnerkolben
Spinner flasks promotes the optimal growth of suspension and microcarrier culture.
  • Impeller height adjusts through compression fitting
  • Indented bottom (up to 1 L) prevents accumulation of microcarrier beads under impeller
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge