Spinner flasks, adjustable hanging bar
Lieferant: BELLCO GLASS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1967-00025 1967-00100A 1967-00250 1967-00500 1967-01000 1967-03000
BELC1967-00025EA 456 EUR InStock
BELC1967-00025 BELC1967-00100A BELC1967-00250 BELC1967-00500 BELC1967-01000 BELC1967-03000
Spinner flasks, adjustable hanging bar
Kolben Spinnerkolben
Spinner flasks, made of borosilicate glass,with PTFE impeller assembly.
  • Height adjustable impeller
  • Impeller assembly rides on glass bearing for friction free operation
  • Flat bottom
The hanging bar spinner flasks have black phenolic rubber-lined central and sidearm caps for 25 and 50 ml flasks.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge