Anaerobic culture tube, Hungate type
Lieferant: BELLCO GLASS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2047-16125
BELC2047-16125EA 1750 EUR InStock
BELC2047-16125
Anaerobic culture tube, Hungate type
Röhrchen Kulturröhrchen
These tubes made of type 1 class B borosilicate glass for growing and maintaining anaerobic bacteria, culture condition.
  • Very good temperature resistance
  • Very good chemical resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge