Screwthread joints
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SQ13UB SQ18UB SQ24UB SQ28UB
BIBBSQ13UBEA 57.7 EUR InStock
BIBBSQ13UB BIBBSQ18UB BIBBSQ24UB BIBBSQ28UB
Screwthread joints
Adapter Adapter, Gewinderohre
Pyrex®, borosilicate glass.
  • Resistance to thermal shock and chemical corrosion
  • Designed to use with the range of plastic screwcaps, silicone rubber rings and PTFE washers
  • Unbadged
It is used by glassblowers in the construction of laboratory glassware.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge