Stopcocks, straight bore
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SSD102 SSD103 SSD104
BIBBSSD102EA 17.3 EUR InStock
BIBBSSD102 BIBBSSD103 BIBBSSD104
Stopcocks, straight bore
Hähne
Straight bore stopcock, made of borosilicat glass 3.3, with plain arm and interchangeable glass key.
  • Glass stopcocks which meet the specifications of DIN 12541
  • Leakage rate meets the requirements of DIN 12540
Lieferumfang: Supplied complete with key retaining device.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge