Alcohol-based rapid disinfectant, ready to use, deconex® SOLARSEPT
Lieferant: Binder
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1002-0022
BIND1002-0022EA 106 EUR InStock
BIND1002-0022
Alcohol-based rapid disinfectant, ready to use, deconex® SOLARSEPT
Reinigungsmittel Desinfektionsmittel
deconex® SOLARSEPT is used for disinfection when a short exposure and drying time is required.
  • aldehyde-free
  • Ready-to-use, with rapid effect ime and short drying time
  • Can also be sprayed on
  • Broad spectrum of activity including tuberculocidal effect
  • Pleasant fresh smell
Binder recommend alcohol based solutions for exterior surfaces, inner chamber, racks and door gaskets.
Wipe disinfection: Pour the product out of the dosing bottle onto a clean cloth or directly onto the surface to be treated (approx. 25 ml product/m²).
Gently wipe the surface all over with steady movements. Makesure that the surface is completely wetted.
Spray disinfection: Spray the surface to be treated, ensuring in particular that it is completely wetted.
Zertifizierungen: Certified by VAH (German Association for Applied Hygiene)
Achtung! Bitte beachten: Due to the alcohol content, spraying is limited for safety reasons to small areas which cannot be reached by wiping.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge