Human skeleton replica
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1000025
BINH1000025EA 0 EUR InStock
BINH1000025
Human skeleton replica
Modelle Humanbiologie
The replica of the human dis-articulated skeleton includes one hand and foot on wire, and one hand and foot loosely articulated.
  • Skull's calvarium sectioned for a closeup view, and the jaw is hinged
  • Sternum's intracostal cartilage is attached
  • Every other bone is life-sized and separated
This disarticulated skeleton is great for any study of the anatomy of the human skeleton.
Lieferumfang: Supplied in a sturdy partitioned cardboard storage box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge