Painted human skull model, 3 parts
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1000055
BINH1000055EA 0 EUR InStock
BINH1000055
Painted human skull model, 3 parts
Modelle Humanbiologie
The muscle origins (red) and insertions (blue) are shown in color on the left side of the skull. Cranial bones and structures are numbered on the right side. This skull model identifies over 140 anatomical details:
  • High-quality original casts
  • Handmade of hard, unbreakable plastic
  • Highly accurate representation of the fissures, foramina, processes, sutures
  • Can be disassembled into skull cap, base of skull and mandible
  • As an option, you can insert a five-part brain (C 18) into the skull
This replica of the human skull is designed for teaching and learning the anatomy of the skull.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge