Venous access device model
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1005563
BINH1005563EA 2110 EUR InStock
BINH1005563
Venous access device model
Modelle Humanbiologie
Teach patients and healthcare staff the care and use of two different types of catheters with this model. Features left and right subclavian catheter connections that may be flushed with water; implanted venous access system with a 'skin' flap over the implant to allow viewing.
  • Mounted on a collapsible easel
  • Made of skinned and lightweight urethane foam
  • May be washed with soap, water as well as antiseptics
Verpackung: In its carrying case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge