Tube holder S
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1014525
BINH1014525EA 1190 EUR InStock
BINH1014525
Tube holder S
Lehrmittel Physik-Lehrmittel Lehrmittel für Elektrizität / Magnetismus
Tube holder to support all S series electron tubes for easy and safe operation. The five pin sockets for the tube are concealed inside the tube holder. A cathode protection switch is integrated into the tube holder, to protect the heated cathode from excessive voltage. The base plate has a slot for attaching the Helmholtz pair of coils S.
  • Terminals : 4 mm safety sockets
  • Dimensions: approx. 130×190×250 mm³
  • Weight: approx. 570 g
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge