Digital counter
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8533341-230
BINH8533341-230EA 0 EUR InStock
BINH8533341-230
Digital counter
Lehrmittel Physik-Lehrmittel Mechanics Learning Activities
Digital counter/timer for measuring duration of motion, transition times, periods, pendulum periods, frequencies, as well as for counting events or Geiger tube pulses. Includes a speaker that can be turned on and off and power supplies for direct connection to photo gate or a Geiger-Müller counter.
  • A fixed counting period ( for events) can be programmed in a range from 0,1 ms to 99999 s
  • Counter events (e.g. start or stop) can either be triggered by a signal to the input sockets or manually via switches
  • Frequency range: 1 to 100 kHz, where voltage >1,5 V
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge