Filter papers for breweries
Lieferant: Ahlstrom-Munksjö
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
120001 120054 146279 146282 120050 120047 120048 120049 120051 120052 120053 120076
BINZ120001EA 6.5 EUR InStock
BINZ120001 BINZ120054 BINZ146279 BINZ146282 BINZ120050 BINZ120047 BINZ120048 BINZ120049 BINZ120051 BINZ120052 BINZ120053 BINZ120076
Filter papers for breweries
Filter Filterpapiere Qualitatives Filterpapier
Ahlstrom-Munksjö offers filters for malt analysis and malt filtration according to MEBAK, ASBC and EBC standards.
Grade 41b: Medium fast filtering. Recommended for malt analysis.
Grade 15: Plain, for final attenuation of congress wort.
Grade 470: Plain kieselguhr paper used for fine turbidity.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge