Lens cleaning tissues
Lieferant: Ahlstrom-Munksjö
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
146099 146100 146102 146103 146104
BINZ146099EA 2.8 EUR InStock
BINZ146099 BINZ146100 BINZ146102 BINZ146103 BINZ146104
Lens cleaning tissues
Wischtücher Wisch- und Handtücher, Einweg Linsenreinigungstücher
These very fine lens cleaning tissues are made of pure manila vegetable fibre.
  • Non abrasive
Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.
12 g/m²
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge