Avantor® ACE® UltraCore BIO, Analytical HPLC/UHPLC Columns
Lieferant: HICHROM - OEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BIO-250-521GEA 637 EUR InStock
BIO-250-521G BIO-250-0521 BIO-250-1021 BIO-250-1521 BIO-250-530G BIO-250-0530 BIO-250-1030 BIO-250-1530 BIO-250-546G BIO-250-0546 BIO-250-1046 BIO-250-1546 BIO-350-521G BIO-350-0521 BIO-350-1021 BIO-350-1521 BIO-350-530G BIO-350-0530 BIO-350-1030 BIO-350-1530 BIO-350-546G BIO-350-0546 BIO-350-1046 BIO-350-1546
Avantor® ACE® UltraCore BIO, Analytical HPLC/UHPLC Columns
Chromatographiesäulen
Avantor® ACE® UltraCore BIO C18 are high performing solid-core (core-shell) particle columns designed for a wide range of biomolecule applications. With both 300 Å and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of more hydrophilic proteins over 5 kDa molecular weight.
  • Wide pore, solid-core combination for excellent peak shape & resolution of large molecules
  • High purity, base deactivated silica with excellent reproducibility
  • Good choice for fast analysis of polypeptides
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge