Human IGFBP7 ELISA kit
Lieferant: BIORBYT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ORB138093-96
BIRBORB138093-96EA 0 EUR InStock
BIRBORB138093-96
Human IGFBP7 ELISA kit
Tests ELISAs
Human IGFBP7 ELISA kit for the quantitative detection of human insulin-like growth factor-binding protein 7 (IGFBP7) in cell culture supernates, serum, plasma (heparin, EDTA) and urine.
Insulin-like growth factor-binding protein 7 is a protein that in humans is encoded by the IGFBP7 gene. The gene was mapped to chromosome 4q12. The major function of the protein is the regulation of availability of insulin-like growth factors (IGFs) in tissue as well as in modulating IGF binding to its receptors. IGFBP7 binds to IGF with high affinity. It also stimulates cell adhesion. The protein is implicated in some cancers.
Lieferumfang: Kit contains 96-well plate precoated with anti- human IGFBP7 antibody, lyophilised recombinant human IGFBP7 standard (2×40 ng tubes), biotinylated anti- human IGFBP7 antibody (130 µl), avidin-biotin-peroxidase complex (130 µl), sample diluent buffer (30 ml), antibody diluent buffer (12 ml), Avidin-Biotin-Peroxidase Complex diluent buffer (12 ml), TMB color developing agent (10 ml), TMB stop solution (10 ml).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge