1,2-Dipalmitoyl-sn-glycero-3-phosphocholine 95%
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
|
|
BD53403-250MG
BLDPBD53403-250MGEA
42
EUR
BLDPBD53403-250MG
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine 95%
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C
Formel:
C₄₀H₈₀NO₈P MW: 734.05 g/mol Storage Temperature: Gefrierschrank |
MDL Number:
MFCD00036903 CAS-Nummer: 63-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...