Mouse recombinant IL-33 (from E. coli)
Lieferant: BIOLEGEND INC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
580509
BLEG580509EA 144 EUR InStock
BLEG580509
Mouse recombinant IL-33 (from E. coli)
Proteine und Peptide
ELISA Standard four-Pack contains four vials of recombinant mouse IL-33 at >5 ng per vial. Recommended for ELISA application only. Standards are calibrated against a BioLegend Master Calibrator.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge