Wire baskets, rectangular, with handle
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10120
BOCH10120EA 272 EUR InStock
BOCH10120
Wire baskets, rectangular, with handle
Körbe Drahtkörbe
These rectangular stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Two cross-ties
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge