Burette and condenser clamps, with swivelling bosshead
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5630
BOCH5630EA 34.7 EUR InStock
BOCH5630
Burette and condenser clamps, with swivelling bosshead
Klemmen Klemmen für den allgemeinen Gebrauch
Burette clamps made of chromium plated steel.
  • Fingers are coated with PVC to protect clamped items
  • With swivelling bosshead (16,5 mm)
  • Angle: 0 to 360°
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge