Burette and test tube clamps, with bosshead
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5631
BOCH5631EA 25.3 EUR InStock
BOCH5631
Burette and test tube clamps, with bosshead
Klemmen Klemmen für den allgemeinen Gebrauch
These chromium plated clamps with bosshead (13,5 mm) and PVC coated jaws are designed for holding burettes and test tubes (Ø 7 - 25 mm).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge