Bossheads, TECHNO
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5670
BOCH5670EA 53 EUR InStock
BOCH5670
Bossheads, TECHNO
Klemmen Stativmuffen
These bossheads are ideal for supporting overhead stirrers.
  • Angle: 90°
  • Thread: M8
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge