Test tube stands, round
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8230 8231 8233
BOCH8230EA 66.1 EUR InStock
BOCH8230 BOCH8231 BOCH8233
Test tube stands, round
Gestelle Reagenzglasgestelle
Rack, made of stainless steel 18/10.
  • With handle
  • Very good tempearture resistance
  • Good chemical resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge