Test tube stands, detachable
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8242 8241 8240
BOCH8242EA 168 EUR InStock
BOCH8242 BOCH8241 BOCH8240
Test tube stands, detachable
Gestelle Reagenzglasgestelle
Made of stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
These test tube stands have detachable frame that saves space while transportation and are available in different dimensions to accommodate various test tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge