Instrument trays
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8484 8486 8480 8485
BOCH8484EA 79.7 EUR InStock
BOCH8484 BOCH8486 BOCH8480 BOCH8485
Instrument trays
Schalen/Schüsseln/Töpfe Instrumentenschalen
Made of 18/10 steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Sturdy
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge