Hand disinfectant, Sterillium® Gel pure
Lieferant: BODE CHEMIE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9817470
BODE9817470EA 0 EUR InStock
BODE9817470
Hand disinfectant, Sterillium® Gel pure
Hautpflege Desinfektionsmittel für die Haut Desinfektionsgel für die Haut
Sterillium Gel pure is suitable for hygienic and surgical rub-in hand disinfection for particularly sensitive skin.
  • Comprehensive antimicrobial activity against bacteria, fungi and viruses
  • Including norovirus activity within application time for hygienic hand disinfection
  • Increases skin hydration with regular use
  • Excellent skin tolerability even with long-term use
  • Possesses an excellent immediate effect
  • Colourant- and fragrance-free
It is ideally suited for hygienic hand disinfection as an alternative to liquid hand disinfectants. It offers comprehensive and rapid efficacy thanks to its highly efficient formula with 85% ethanol. It evaporates quickly and leaves hands feeling soft - not sticky.
Sterillium Gel pure is used as ready-to-use alcohol-based rub-in product that is independently of water and wash-basin to prevent infection in all areas of health care and industry where hygiene is important as well as in home dialysis and when travelling.
Zertifizierungen: * tested with the murine norovirus (MNV)
Achtung! Bitte beachten: Use biocides safely. Always read the label and product information before use.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge