Microwave digestion vessels, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A240-02 A240-06 A240-08 A240-10 A244-06 A244-04 A244-02
BOHLA240-02EA 396 EUR InStock
BOHLA240-02 BOHLA240-06 BOHLA240-08 BOHLA240-10 BOHLA244-06 BOHLA244-04 BOHLA244-02
Microwave digestion vessels, BOLA
Aufschlusssysteme Aufschluss-Systeme
Dimensionally stable basic vessel and screw cover made of TFM, homogenous, non porous surface.
  • One piece of sealing and rupture membranes already mounted
  • For samples of up to maximum 0,5 g
The liner allows a more precise weighted sample and different digestions with only one basic vessel.
Lieferumfang: Includes 10 pieces of replacement membranes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge