Stirrer bearings, PTFE, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B288-02
BOHLB288-02EA 81.2 EUR InStock
BOHLB288-02
Stirrer bearings, PTFE, BOLA
Rührer Rührwerkzeuge
PTFE stirrer bearings with adjustable special gasket.
  • Chemical resistance
  • Temperature resistance: −20 to 250 °C
Integrated special nut is present for easy locking and unlocking of the ground joint.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge