Ground joint tube fittings, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B304-24 B304-22 B304-20 B304-16 B304-10
BOHLB304-24EA 62.3 EUR InStock
BOHLB304-24 BOHLB304-22 BOHLB304-20 BOHLB304-16 BOHLB304-10
Ground joint tube fittings, BOLA
Adapter Adapter, Normschliff mit Schlaucholiven
PTFE, white.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
For connecting tubes, hard-walled tubing, thermometers. The sealing rings on the outside of the cone prevent sticking of the ground joints and improve the sealing. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge