Magnetic Stirring Bars, PTFE, cylindrical, with pivot ring, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C354-05 C354-11 C354-08 C354-23 C354-35 C354-32 C354-29 C354-26 C354-20 C354-17 C354-14 C354-02
BOHLC354-05EA 2 EUR InStock
BOHLC354-05 442-9837 442-9836 442-9838 BOHLC354-35 BOHLC354-32 BOHLC354-29 BOHLC354-26 BOHLC354-20 BOHLC354-17 BOHLC354-14 BOHLC354-02
Magnetic Stirring Bars, PTFE, cylindrical, with pivot ring, BOLA
Rührer Magnetrührstäbchen
PTFE coated stirring bars. Provide excellent centering and smooth stirring.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge