Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Magnetic Stirring Bars, PTFE, BOLA

Lieferant: Bohlender

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C362-08 C362-16 C362-20 C362-12 C362-05 C362-04 C362-14 C362-02 C362-07 C362-24
BOHLC362-08EA 119 EUR InStock
BOHLC362-08 BOHLC362-16 BOHLC362-20 BOHLC362-12 BOHLC362-05 BOHLC362-04 BOHLC362-14 BOHLC362-02 BOHLC362-07 BOHLC362-24
Magnetic Stirring Bars, PTFE, BOLA
Rührer Magnetrührstäbchen
PTFE encapsulated magnetic core (Alnico 5) axially mounted in a guide cage made of PTFE.

  • Universal chemical resistance
  • Autoclavable at 121 °C
  • Temperature resistance: –200 to +250 °C

This magnetic stirring bar is mounted in a guide cage which prevents shear action on the bottom of the beaker and the liquid is mixed carefully. This cage can easily be inserted or removed.

Optimum mixing results and smooth running in glass beakers also on an uneven bottom.

Zertifizierungen: Conforms to FDA.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge