Overhead stirrer tools, impellers, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C389-28 C389-32 C389-66 C389-20 C389-18 C389-62 C389-36 C389-22
BOHLC389-28EA 328 EUR InStock
BOHLC389-28 BOHLC389-32 BOHLC389-66 BOHLC389-20 BOHLC389-18 BOHLC389-62 BOHLC389-36 BOHLC389-22
Overhead stirrer tools, impellers, BOLA
Rührer Rührwerkzeuge
Stirrer shafts made of PTFE-jacketed stainless steel and the impeller completely made of PTFE with three blades bent backwards.
  • Suitable for stirring in vessels with round bottom
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge