Stirrer bearings, PTFE
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C424-04 C424-05 C424-08 C424-09 C424-12 C424-13 C424-14 C424-16 C424-18 C429-18 C429-14
BOHLC424-04EA 42.9 EUR InStock
BOHLC424-04 BOHLC424-05 BOHLC424-08 BOHLC424-09 BOHLC424-12 BOHLC424-13 BOHLC424-14 BOHLC424-16 BOHLC424-18 BOHLC429-18 BOHLC429-14
Stirrer bearings, PTFE
Rührer Rührwerkzeuge
These bearings have sealing rings to ensure a perfect seal. A special gasket made of PTFE and a FPM o-ring which is compressed by a GL screw cap ensure the stirrer shaft is sealed.
  • Cone can be removed easily from the socket
  • Ground joint does not stick and danger of breaking is reduced
  • Gasket can be exchanged when worn
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge