Overhead stirrer tools, radial stirrers, six paddles, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C598-12 C598-26 C598-32 C598-22 C598-16 C598-46 C598-36 C598-42 C598-56 C598-18
BOHLC598-12EA 161 EUR InStock
BOHLC598-12 BOHLC598-26 BOHLC598-32 BOHLC598-22 BOHLC598-16 BOHLC598-46 BOHLC598-36 BOHLC598-42 BOHLC598-56 BOHLC598-18
Overhead stirrer tools, radial stirrers, six paddles, BOLA
Rührer Rührwerkzeuge
Stirrer shafts made of PTFE-jacketed stainless steel and the impeller completely made of PTFE with six radial paddles.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Axial suction of mixture, strong radial flow. Ideal for aerating liquids.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge