Socket GL tube fittings, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D571-32 D571-16 D571-08
BOHLD571-32EA 45.5 EUR InStock
BOHLD571-32 BOHLD571-16 BOHLD571-08
Socket GL tube fittings, BOLA
Adapter Übergangsstücke mit Kegelschliffen
PTFE, white.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Vacuum: suitable
Fitting with socket for tubes with ground joint and GL thread for connection of hard-walled tubing (e.g. PTFE, PFA, FEP) with laboratory screw joints. The products is only exposed to PTFE.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge