Distributors for reaction vessels (S), BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D748-16 D748-60 D748-40
BOHLD748-16EA 249 EUR InStock
BOHLD748-16 BOHLD748-60 BOHLD748-40
Distributors for reaction vessels (S), BOLA
Verschlüsse Flaschenverschlüsse
PP screw cap, blue and PTFE body.
  • Very good chemical reistance
  • For working temperatures up to max. +250 °C (pp screw cap max. +140 °C)
  • FDC conform
Screw cap with GLS 80® thread, body with center ground joint and lateral GL-threaded or ground joint necks. Applications: Drawing or inserting aggressive or pure liquids. Inserting tubing, tubes and probes into vessels. Use of a stirrer bearing in center neck for centrical position of a stirrer shaft.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge